Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTTTTTGGTCTGTGTAGTAGGAA[A/G]GGGTATTGAGGGAGGCCACCTTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608924 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FOXP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FOXP4 - forkhead box P4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012426.1 | Intron | NP_001012426.1 | ||||
NM_001012427.1 | Intron | NP_001012427.1 | ||||
NM_138457.2 | Intron | NP_612466.1 | ||||
XM_006714991.3 | Intron | XP_006715054.1 | ||||
XM_011514289.2 | Intron | XP_011512591.1 | ||||
XM_011514290.2 | Intron | XP_011512592.1 | ||||
XM_011514291.2 | Intron | XP_011512593.1 | ||||
XM_011514292.2 | Intron | XP_011512594.1 | ||||
XM_011514293.2 | Intron | XP_011512595.1 | ||||
XM_017010233.1 | Intron | XP_016865722.1 | ||||
XM_017010234.1 | Intron | XP_016865723.1 |
FOXP4-AS1 - FOXP4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |