Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAATGGGAGAAAGCAGCGAGTGA[C/G]AGGGGAAGGGGCGCCAGGCGAGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604971 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MDFI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MDFI - MyoD family inhibitor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300804.1 | 69 | Intron | NP_001287733.1 | |||
NM_001300805.1 | 69 | UTR 5 | NP_001287734.1 | |||
NM_001300806.1 | 69 | UTR 5 | NP_001287735.1 | |||
NM_005586.3 | 69 | UTR 5 | NP_005577.1 | |||
XM_005249117.3 | 69 | UTR 5 | XP_005249174.1 | |||
XM_011514625.2 | 69 | Intron | XP_011512927.1 | |||
XM_011514626.2 | 69 | Intron | XP_011512928.1 | |||
XM_017010867.1 | 69 | Intron | XP_016866356.1 |