Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAACAGCATTTGGGCATGCCAAAG[A/G]AAGTAGCTGCTCCCTTCTTCTGCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612915 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED20 - mediator complex subunit 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305455.1 | Intron | NP_001292384.1 | ||||
NM_001305456.1 | Intron | NP_001292385.1 | ||||
NM_001305457.1 | Intron | NP_001292386.1 | ||||
NM_004275.4 | Intron | NP_004266.2 |