Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCGAGCTAGCCACCAGCATGCT[A/G]TACTCCCCAGGTAGGCACTCATGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604822 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPN11 - calpain 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007058.3 | 44 | Silent Mutation | CTA,CTG | L,L 2 | NP_008989.2 | |
XM_006714984.1 | 44 | Silent Mutation | CTA,CTG | L,L 2 | XP_006715047.1 | |
XM_006714985.1 | 44 | Silent Mutation | CTA,CTG | L,L 2 | XP_006715048.1 | |
XM_006714987.1 | 44 | Silent Mutation | CTA,CTG | L,L 2 | XP_006715050.1 | |
XM_011514274.1 | 44 | Intron | XP_011512576.1 | |||
XM_011514275.1 | 44 | UTR 5 | XP_011512577.1 |
TMEM63B - transmembrane protein 63B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |