Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATCAATCAACATACATTTATTG[A/G]ATATACTCTGTAAAGAAGGTGCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605994 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP39A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP39A1 - cytochrome P450 family 39 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278738.1 | 2085 | UTR 3 | NP_001265667.1 | |||
NM_001278739.1 | 2085 | UTR 3 | NP_001265668.1 | |||
NM_016593.4 | 2085 | UTR 3 | NP_057677.2 | |||
XM_005249171.3 | 2085 | Intron | XP_005249228.1 | |||
XM_011514658.1 | 2085 | UTR 3 | XP_011512960.1 | |||
XM_017010921.1 | 2085 | Intron | XP_016866410.1 | |||
XM_017010922.1 | 2085 | Intron | XP_016866411.1 | |||
XM_017010923.1 | 2085 | Intron | XP_016866412.1 | |||
XM_017010924.1 | 2085 | UTR 3 | XP_016866413.1 | |||
XM_017010925.1 | 2085 | UTR 3 | XP_016866414.1 |