Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACTAACAATAACATGTTTAAGG[A/G]TGTCATTTGTTTCACATCTCAAACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616908 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTCHD4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTCHD4 - patched domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013732.3 | Intron | NP_001013754.3 | ||||
NM_207499.2 | Intron | NP_997382.2 | ||||
XM_011514639.2 | Intron | XP_011512941.1 | ||||
XM_017010891.1 | Intron | XP_016866380.1 | ||||
XM_017010892.1 | Intron | XP_016866381.1 | ||||
XM_017010893.1 | Intron | XP_016866382.1 | ||||
XM_017010894.1 | Intron | XP_016866383.1 | ||||
XM_017010895.1 | Intron | XP_016866384.1 | ||||
XM_017010896.1 | Intron | XP_016866385.1 |