Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTACAGTTACTATAGAGATCTT[C/T]GTACTTGCAACCATTGGCTGGAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CRISP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRISP3 - cysteine rich secretory protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190986.1 | 755 | Missense Mutation | AAA,GAA | K,E 233 | NP_001177915.1 | |
NM_006061.2 | 755 | Missense Mutation | AAA,GAA | K,E 223 | NP_006052.2 |