Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAACTTTAGTCGTTATAACAACTG[C/T]GACTGTTGAGAAATTTCACTGTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609091 MIM: 612325 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO9 - F-box protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012347.4 | 48 | Intron | NP_036479.1 | |||
NM_033480.2 | 48 | Intron | NP_258441.1 | |||
NM_033481.3 | 48 | UTR 5 | NP_258442.2 | |||
XM_005248995.4 | 48 | Intron | XP_005249052.1 | |||
XM_005248996.4 | 48 | Intron | XP_005249053.1 |
ICK - intestinal cell (MAK-like) kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105375092 - uncharacterized LOC105375092 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |