Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATAAACTACTGACAATATGTACAC[A/T]GTGTTAAGCCTGCTTGATTAAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGCLL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMGCLL1 - 3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042406.1 | 1443 | UTR 3 | NP_001035865.1 | |||
NM_001287741.1 | 1443 | UTR 3 | NP_001274670.1 | |||
NM_001287746.1 | 1443 | UTR 3 | NP_001274675.1 | |||
NM_001287753.1 | 1443 | UTR 3 | NP_001274682.1 | |||
NM_019036.2 | 1443 | UTR 3 | NP_061909.2 | |||
XM_011514700.2 | 1443 | Intron | XP_011513002.1 |