Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGCTCAGCTCCTAGACCCACCCC[C/T]ACAACCTTCATCCAACCTGTCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NRN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NRN1 - neuritin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278710.1 | Intron | NP_001265639.1 | ||||
NM_001278711.1 | Intron | NP_001265640.1 | ||||
NM_016588.2 | Intron | NP_057672.1 | ||||
XM_006715106.3 | Intron | XP_006715169.1 | ||||
XM_006715107.3 | Intron | XP_006715170.1 |