Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACCCAGCTTTGATGGTGGGGC[A/G]ACTCCCGTGTCTGCAGCTGTGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610699 | ||||||||||||||||||||
Literature Links: |
LOC101927021 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC101927021 - uncharacterized LOC101927021 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZFAND2A - zinc finger AN1-type containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182491.2 | 716 | Missense Mutation | CGC,TGC | R,C 139 | NP_872297.2 | |
XM_017012774.1 | 716 | Missense Mutation | CGC,TGC | R,C 139 | XP_016868263.1 |