Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGGCTCGGCGCCCTCACCTCCT[C/T]GGACAGACCAGCAACGCCCACGCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611345 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
INTS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INTS1 - integrator complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080453.2 | 6925 | UTR 3 | NP_001073922.2 | |||
XM_011515260.1 | 6925 | UTR 3 | XP_011513562.1 | |||
XM_011515262.2 | 6925 | Intron | XP_011513564.1 | |||
XM_017011959.1 | 6925 | UTR 3 | XP_016867448.1 | |||
XM_017011960.1 | 6925 | UTR 3 | XP_016867449.1 |