Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACAGTAAATATTTTAATTTACCT[A/G]TGGTTCAACAAGTAACTCATAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM196 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM196 - transmembrane protein 196 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152774.3 | 2211 | UTR 3 | NP_689987.3 | |||
XM_005249692.4 | 2211 | Intron | XP_005249749.2 | |||
XM_011515249.2 | 2211 | Intron | XP_011513551.1 | |||
XM_011515250.2 | 2211 | Intron | XP_011513552.1 | |||
XM_017011928.1 | 2211 | Intron | XP_016867417.1 | |||
XM_017011929.1 | 2211 | Intron | XP_016867418.1 |