Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCGCCAAGCACAGCCCTAAAACAA[C/T]TCTCGTCCACTTTCCTGTTAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612927 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AVL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AVL9 - AVL9 cell migration associated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015060.2 | 5118 | UTR 3 | NP_055875.1 | |||
XM_005249668.2 | 5118 | UTR 3 | XP_005249725.1 | |||
XM_005249669.2 | 5118 | UTR 3 | XP_005249726.1 | |||
XM_005249670.2 | 5118 | UTR 3 | XP_005249727.1 | |||
XM_005249671.4 | 5118 | Intron | XP_005249728.1 | |||
XM_011515227.2 | 5118 | UTR 3 | XP_011513529.1 | |||
XM_017011891.1 | 5118 | UTR 3 | XP_016867380.1 | |||
XM_017011892.1 | 5118 | UTR 3 | XP_016867381.1 |
DPY19L1P1 - DPY19L1 pseudogene 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |