Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATTGGCGGCGCCCCACTACTGCT[A/G]GTGGTTCCTGGGATTAATGCCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KBTBD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KBTBD2 - kelch repeat and BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015483.2 | 3567 | UTR 3 | NP_056298.2 | |||
XM_005249696.2 | 3567 | UTR 3 | XP_005249753.1 | |||
XM_005249697.2 | 3567 | Intron | XP_005249754.1 | |||
XM_011515256.2 | 3567 | UTR 3 | XP_011513558.1 | |||
XM_011515257.2 | 3567 | Intron | XP_011513559.1 |