Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATGTGAAAATGAAGGTGAAGT[C/T]CTCCAGATTCCATTTATCACAGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608699 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMPER PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMPER - BMP binding endothelial regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133468.4 | 202 | Silent Mutation | GTC,GTT | V,V 56 | NP_597725.1 | |
XM_005249633.2 | 202 | Silent Mutation | GTC,GTT | V,V 56 | XP_005249690.1 | |
XM_017011800.1 | 202 | UTR 5 | XP_016867289.1 | |||
XM_017011801.1 | 202 | UTR 5 | XP_016867290.1 |