Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAATGGGGTTCCTTCTACTACT[A/C]CTTTAAGGTAAGTTTCTTGCCTGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608595 MIM: 608596 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPSR1 - neuropeptide S receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300933.1 | 314 | Missense Mutation | TAC,TCC | Y,S 47 | NP_001287862.1 | |
NM_001300934.1 | 314 | Missense Mutation | TAC,TCC | Y,S 47 | NP_001287863.1 | |
NM_001300935.1 | 314 | Missense Mutation | TAC,TCC | Y,S 47 | NP_001287864.1 | |
NM_207172.1 | 314 | Missense Mutation | TAC,TCC | Y,S 47 | NP_997055.1 | |
NM_207173.1 | 314 | Missense Mutation | TAC,TCC | Y,S 47 | NP_997056.1 |
NPSR1-AS1 - NPSR1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |