Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTCCTATTTCTCTTTAATTTAG[A/G]ATACATAATGATTTGTGATTTGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIAA0895 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KIAA0895 - KIAA0895 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001100425.1 | Intron | NP_001093895.1 | ||||
NM_001199706.1 | Intron | NP_001186635.1 | ||||
NM_001199707.1 | Intron | NP_001186636.1 | ||||
NM_001199708.1 | Intron | NP_001186637.1 | ||||
NM_001300956.1 | Intron | NP_001287885.1 | ||||
NM_015314.2 | Intron | NP_056129.2 | ||||
XM_005249689.3 | Intron | XP_005249746.1 | ||||
XM_005249690.4 | Intron | XP_005249747.1 |