Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGCTTACTTACAGGTTGATCA[A/G]TTGTAGGAAAGAATACCTATGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609187 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUGCT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SUGCT - succinyl-CoA:glutarate-CoA transferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193311.1 | Intron | NP_001180240.1 | ||||
NM_001193312.1 | Intron | NP_001180241.1 | ||||
NM_001193313.1 | Intron | NP_001180242.1 | ||||
NM_024728.2 | Intron | NP_079004.1 | ||||
XM_006715775.3 | Intron | XP_006715838.1 | ||||
XM_011515525.2 | Intron | XP_011513827.1 | ||||
XM_011515526.2 | Intron | XP_011513828.1 | ||||
XM_011515527.2 | Intron | XP_011513829.1 | ||||
XM_011515528.2 | Intron | XP_011513830.1 | ||||
XM_011515529.2 | Intron | XP_011513831.1 | ||||
XM_011515530.2 | Intron | XP_011513832.1 | ||||
XM_017012621.1 | Intron | XP_016868110.1 | ||||
XM_017012622.1 | Intron | XP_016868111.1 |