Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGAACAGATAAAAGTAGCAATGT[G/T]GCTGAGTGTCACCAACTGTCCGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 165240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLI3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLI3 - GLI family zinc finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000168.5 | 5681 | UTR 3 | NP_000159.3 | |||
XM_011515274.2 | 5681 | Intron | XP_011513576.1 | |||
XM_017011997.1 | 5681 | Intron | XP_016867486.1 |