Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATGTTGGTGGTGCTCATGGACAG[A/C]GCTGGGAATGGGAGGGACGCCCGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 165240 | ||||||||||||||||||||
Literature Links: |
GLI3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLI3 - GLI family zinc finger 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000168.5 | 4738 | Silent Mutation | GCG,GCT | A,A 1549 | NP_000159.3 | |
XM_011515274.2 | 4738 | Silent Mutation | GCG,GCT | A,A 1490 | XP_011513576.1 | |
XM_017011997.1 | 4738 | Silent Mutation | GCG,GCT | A,A 1548 | XP_016867486.1 |