Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCCACCAGACAGCGTGGCTCAA[C/T]GGGAGCAAGGCGCGCAGGGACAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610296 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUDCD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUDCD3 - NudC domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015332.3 | 3425 | UTR 3 | NP_056147.2 | |||
XM_011515247.2 | 3425 | Intron | XP_011513549.1 | |||
XM_017011908.1 | 3425 | Intron | XP_016867397.1 | |||
XM_017011909.1 | 3425 | UTR 3 | XP_016867398.1 | |||
XM_017011910.1 | 3425 | UTR 3 | XP_016867399.1 |