Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAAACTTCAGGACAAAAATGTTT[C/T]ATTTAAGGACTTGTGCTGCTAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613022 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
OGDH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OGDH - oxoglutarate dehydrogenase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003941.2 | 98 | Missense Mutation | CAT,TAT | H,Y 3 | NP_001003941.1 | |
NM_001165036.1 | 98 | Missense Mutation | CAT,TAT | H,Y 3 | NP_001158508.1 | |
NM_002541.3 | 98 | Missense Mutation | CAT,TAT | H,Y 3 | NP_002532.2 | |
XM_005249759.4 | 98 | Missense Mutation | CAT,TAT | H,Y 3 | XP_005249816.1 | |
XM_005249761.3 | 98 | Missense Mutation | CAT,TAT | H,Y 3 | XP_005249818.1 | |
XM_011515408.2 | 98 | Missense Mutation | CAT,TAT | H,Y 3 | XP_011513710.1 |