Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCAAGGCAAGCTGCTTGTATCA[A/G]TAGCAAAACCCTGACTGAAGAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608191 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RBAK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RBAK - RB associated KRAB zinc finger | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204456.1 | 641 | Missense Mutation | AAT,AGT | N,S 106 | NP_001191385.1 | |
NM_021163.3 | 641 | Missense Mutation | AAT,AGT | N,S 106 | NP_066986.1 |
RBAK-RBAKDN - RBAK-RBAKDN readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204513.1 | 641 | Intron | NP_001191442.1 |
RBAKDN - RBAK downstream neighbor (non-protein coding) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |