Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACATTCCGATTCCCCACGGCTGAG[C/T]TCATCTGCGTCGTAAGCCGCCCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 173430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDGFA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDGFA - platelet derived growth factor subunit A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002607.5 | Intron | NP_002598.4 | ||||
NM_033023.4 | Intron | NP_148983.1 | ||||
XM_011515415.1 | Intron | XP_011513717.1 | ||||
XM_011515416.1 | Intron | XP_011513718.1 | ||||
XM_011515417.1 | Intron | XP_011513719.1 | ||||
XM_011515418.1 | Intron | XP_011513720.1 | ||||
XM_011515419.2 | Intron | XP_011513721.1 | ||||
XM_017012289.1 | Intron | XP_016867778.1 |