Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAGGCACGTCCTCAGAGCCTTG[G/T]CCCTGGCTTACTTGCCTTTCCTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GIMAP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GIMAP6 - GTPase, IMAP family member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244071.1 | 1024 | UTR 3 | NP_001231000.1 | |||
NM_001244072.1 | 1024 | Silent Mutation | GGA,GGC | G,G 326 | NP_001231001.1 | |
NM_024711.5 | 1024 | Silent Mutation | GGA,GGC | G,G 256 | NP_078987.3 |