Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTTAGGGTGAATTAGGCCTCCT[G/T]CTCCCTACTATAATTTTCTTCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616813 MIM: 614792 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGAP3 - ArfGAP with GTPase domain, ankyrin repeat and PH domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042535.3 | Intron | NP_001036000.1 | ||||
NM_001281300.1 | Intron | NP_001268229.1 | ||||
NM_001308304.1 | Intron | NP_001295233.1 | ||||
NM_001308305.1 | Intron | NP_001295234.1 | ||||
NM_031946.6 | Intron | NP_114152.3 | ||||
XM_005249943.2 | Intron | XP_005250000.1 | ||||
XM_011515780.1 | Intron | XP_011514082.1 | ||||
XM_017011732.1 | Intron | XP_016867221.1 | ||||
XM_017011733.1 | Intron | XP_016867222.1 | ||||
XM_017011734.1 | Intron | XP_016867223.1 | ||||
XM_017011735.1 | Intron | XP_016867224.1 |
TMUB1 - transmembrane and ubiquitin like domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |