Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGAAAACAAACGGCGGCCGCTGC[C/T]GCATCCGGGCACTCTGCTGGTCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607981 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NUB1 - negative regulator of ubiquitin like proteins 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243351.1 | 6 | Missense Mutation | CGC,TGC | R,C 9 | NP_001230280.1 | |
NM_016118.4 | 6 | Missense Mutation | CGC,TGC | R,C 9 | NP_057202.3 | |
XM_005250011.2 | 6 | Intron | XP_005250068.1 | |||
XM_017012304.1 | 6 | Missense Mutation | CGC,TGC | R,C 9 | XP_016867793.1 | |
XM_017012305.1 | 6 | Missense Mutation | CGC,TGC | R,C 9 | XP_016867794.1 | |
XM_017012306.1 | 6 | UTR 5 | XP_016867795.1 | |||
XM_017012307.1 | 6 | Missense Mutation | CGC,TGC | R,C 9 | XP_016867796.1 | |
XM_017012308.1 | 6 | Missense Mutation | CGC,TGC | R,C 9 | XP_016867797.1 | |
XM_017012309.1 | 6 | Intron | XP_016867798.1 | |||
XM_017012310.1 | 6 | Intron | XP_016867799.1 | |||
XM_017012311.1 | 6 | UTR 5 | XP_016867800.1 | |||
XM_017012312.1 | 6 | Intron | XP_016867801.1 | |||
XM_017012313.1 | 6 | Intron | XP_016867802.1 | |||
XM_017012314.1 | 6 | Intron | XP_016867803.1 |