Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTTGATGCTTTTGTGTCTCTG[A/C]CCCTTAGCAATAACCTCCAAAACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603134 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUL1 - cullin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003592.2 | Intron | NP_003583.2 | ||||
XM_005250060.4 | Intron | XP_005250117.1 | ||||
XM_011516629.2 | Intron | XP_011514931.1 | ||||
XM_011516630.2 | Intron | XP_011514932.1 | ||||
XM_011516631.2 | Intron | XP_011514933.1 | ||||
XM_011516632.2 | Intron | XP_011514934.1 | ||||
XM_017012723.1 | Intron | XP_016868212.1 |