Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCAGAAGCCCGCACTGCACCAC[C/T]GTGTTGCCCTTGGAGAGCTCCAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615732 MIM: 612548 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSUN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NSUN5 - NOP2/Sun RNA methyltransferase family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIM50 - tripartite motif containing 50 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281450.1 | 1068 | Silent Mutation | ACA,ACG | T,T 313 | NP_001268379.1 | |
NM_001281451.1 | 1068 | Silent Mutation | ACA,ACG | T,T 314 | NP_001268380.1 | |
NM_178125.3 | 1068 | Silent Mutation | ACA,ACG | T,T 314 | NP_835226.2 | |
XM_011515787.1 | 1068 | UTR 3 | XP_011514089.1 | |||
XM_011515788.2 | 1068 | Silent Mutation | ACA,ACG | T,T 105 | XP_011514090.1 | |
XM_011515789.2 | 1068 | Silent Mutation | ACA,ACG | T,T 313 | XP_011514091.2 |