Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCACCCATTTTTTCCTTGGATGG[A/G]TACAGACTTTCTTGCCTCTTTTGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604697 | ||||||||||||||||||||
Literature Links: |
CCL26 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCL26 - C-C motif chemokine ligand 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006072.4 | 415 | Missense Mutation | ACC,ATC | T,I 74 | NP_006063.1 | |
XM_017011671.1 | 415 | Missense Mutation | ACC,ATC | T,I 128 | XP_016867160.1 | |
XM_017011672.1 | 415 | Missense Mutation | ACC,ATC | T,I 74 | XP_016867161.1 |