Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGACCAGCTCTGTAAAAGCCCC[A/G]CCTCTGCCTTCGGATAGGTAAGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603989 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6839 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR6839 - microRNA 6839 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF107 - zinc finger protein 107 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013746.2 | 786 | Intron | NP_001013768.1 | |||
NM_001282359.1 | 786 | Intron | NP_001269288.1 | |||
NM_001282360.1 | 786 | Intron | NP_001269289.1 | |||
NM_016220.4 | 786 | Intron | NP_057304.1 | |||
XM_017012283.1 | 786 | UTR 5 | XP_016867772.1 | |||
XM_017012284.1 | 786 | UTR 5 | XP_016867773.1 | |||
XM_017012285.1 | 786 | UTR 5 | XP_016867774.1 | |||
XM_017012286.1 | 786 | Intron | XP_016867775.1 |