Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAAAACATACTAGAATATGAAGA[A/T]CCAAAATATGAACCAAAAGCAAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609907 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA3D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SEMA3D - semaphorin 3D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152754.2 | 6005 | UTR 3 | NP_689967.2 | |||
XM_011515960.1 | 6005 | UTR 3 | XP_011514262.1 | |||
XM_011515961.2 | 6005 | UTR 3 | XP_011514263.1 | |||
XM_017011873.1 | 6005 | UTR 3 | XP_016867362.1 |