Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATAAAGATATTTTCCATTCTGAT[A/G]CTTTCAGCCTATTTGACTCAACGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604080 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF138 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF138 - zinc finger protein 138 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160183.2 | Intron | NP_001153655.1 | ||||
NM_001271637.1 | Intron | NP_001258566.1 | ||||
NM_001271638.1 | Intron | NP_001258567.1 | ||||
NM_001271639.1 | Intron | NP_001258568.1 | ||||
NM_001271640.1 | Intron | NP_001258569.1 | ||||
NM_001271649.1 | Intron | NP_001258578.1 | ||||
NM_006524.3 | Intron | NP_006515.2 |