Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCTCTCAAATGAAGTGAATGGA[A/G]CTGAGACTAAATTTCTGTCTGTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611232 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLDN12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLDN12 - claudin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185072.2 | Intron | NP_001172001.1 | ||||
NM_001185073.2 | Intron | NP_001172002.1 | ||||
NM_012129.4 | Intron | NP_036261.1 |
LOC101409256 - cell division cycle 42 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC102723899 - pituitary tumor-transforming gene 1 protein-interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011516764.2 | Intron | XP_011515066.1 | ||||
XM_017012889.1 | Intron | XP_016868378.1 | ||||
XM_017012890.1 | Intron | XP_016868379.1 | ||||
XM_017012891.1 | Intron | XP_016868380.1 | ||||
XM_017012892.1 | Intron | XP_016868381.1 | ||||
XM_017012893.1 | Intron | XP_016868382.1 | ||||
XM_017012894.1 | Intron | XP_016868383.1 |