Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTGACAGCTGTCTCAGCAGTGG[C/T]TTGTGTGTGCCACAAGTGCCAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604001 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKAP9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AKAP9 - A-kinase anchoring protein 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005751.4 | Intron | NP_005742.4 | ||||
NM_147185.2 | Intron | NP_671714.1 | ||||
XM_017011642.1 | Intron | XP_016867131.1 | ||||
XM_017011643.1 | Intron | XP_016867132.1 | ||||
XM_017011644.1 | Intron | XP_016867133.1 | ||||
XM_017011645.1 | Intron | XP_016867134.1 | ||||
XM_017011646.1 | Intron | XP_016867135.1 | ||||
XM_017011647.1 | Intron | XP_016867136.1 | ||||
XM_017011648.1 | Intron | XP_016867137.1 | ||||
XM_017011649.1 | Intron | XP_016867138.1 | ||||
XM_017011650.1 | Intron | XP_016867139.1 | ||||
XM_017011651.1 | Intron | XP_016867140.1 | ||||
XM_017011652.1 | Intron | XP_016867141.1 | ||||
XM_017011653.1 | Intron | XP_016867142.1 | ||||
XM_017011654.1 | Intron | XP_016867143.1 | ||||
XM_017011655.1 | Intron | XP_016867144.1 | ||||
XM_017011656.1 | Intron | XP_016867145.1 | ||||
XM_017011657.1 | Intron | XP_016867146.1 | ||||
XM_017011658.1 | Intron | XP_016867147.1 | ||||
XM_017011659.1 | Intron | XP_016867148.1 | ||||
XM_017011660.1 | Intron | XP_016867149.1 |