Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGGGGAGGGGGAGTGCTCACAC[A/G]GTGTAAGTTTTGACAGACTTATAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TMEM130 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TMEM130 - transmembrane protein 130 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134450.1 | 1149 | Silent Mutation | ACC,ACT | T,T 434 | NP_001127922.1 | |
NM_001134451.1 | 1149 | Silent Mutation | ACC,ACT | T,T 320 | NP_001127923.1 | |
NM_152913.2 | 1149 | Silent Mutation | ACC,ACT | T,T 422 | NP_690877.1 |