Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCAAATGCAACATTCCTGCTTCA[A/G]AGTCTTTTAAACCATGAGACCCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605568 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927550 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927550 - uncharacterized LOC101927550 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMURF1 - SMAD specific E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199847.1 | 4469 | UTR 3 | NP_001186776.1 | |||
NM_020429.2 | 4469 | UTR 3 | NP_065162.1 | |||
NM_181349.2 | 4469 | UTR 3 | NP_851994.1 | |||
XM_017012457.1 | 4469 | UTR 3 | XP_016867946.1 |