Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGACACGGGACACCAACTTCGGGAC[A/G]AACTGCATCTGCCGCGTGCCCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603477 MIM: 607075 MIM: 614774 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5J2-PTCD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5J2-PTCD1 - ATP5J2-PTCD1 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198879.1 | 4688 | UTR 3 | NP_001185808.1 |
BUD31 - BUD31 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003910.3 | 4688 | Silent Mutation | ACA,ACG | T,T 115 | NP_003901.2 | |
XM_005250670.4 | 4688 | Silent Mutation | ACA,ACG | T,T 115 | XP_005250727.1 | |
XM_005250671.4 | 4688 | Silent Mutation | ACA,ACG | T,T 115 | XP_005250728.1 | |
XM_005250674.3 | 4688 | Silent Mutation | ACA,ACG | T,T 115 | XP_005250731.1 | |
XM_017012760.1 | 4688 | Silent Mutation | ACA,ACG | T,T 115 | XP_016868249.1 | |
XM_017012761.1 | 4688 | Silent Mutation | ACA,ACG | T,T 115 | XP_016868250.1 | |
XM_017012762.1 | 4688 | Silent Mutation | ACA,ACG | T,T 115 | XP_016868251.1 |
PDAP1 - PDGFA associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTCD1 - pentatricopeptide repeat domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015545.3 | 4688 | UTR 3 | NP_056360.2 |