Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCCACAAGCCTGATAAAAAATA[C/T]ATGTTGTTGGCAGTTGAAATTGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606821 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COG5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COG5 - component of oligomeric golgi complex 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DUS4L - dihydrouridine synthase 4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270419.1 | Intron | NP_001257348.1 | ||||
NM_181581.2 | Intron | NP_853559.1 | ||||
XM_005250125.4 | Intron | XP_005250182.1 | ||||
XM_006715844.2 | Intron | XP_006715907.1 | ||||
XM_006715845.3 | Intron | XP_006715908.1 | ||||
XM_011515766.2 | Intron | XP_011514068.1 | ||||
XM_011515767.2 | Intron | XP_011514069.1 | ||||
XM_017011708.1 | Intron | XP_016867197.1 | ||||
XM_017011709.1 | Intron | XP_016867198.1 | ||||
XM_017011710.1 | Intron | XP_016867199.1 | ||||
XM_017011711.1 | Intron | XP_016867200.1 | ||||
XM_017011712.1 | Intron | XP_016867201.1 | ||||
XM_017011713.1 | Intron | XP_016867202.1 |