Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAAAAGAGTTCTAACGGGACTAA[C/T]TTTGCTGTTCTTCAGCCCTGTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600833 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ST7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ST7 - suppression of tumorigenicity 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018412.3 | Intron | NP_060882.2 | ||||
NM_021908.2 | Intron | NP_068708.1 |
ST7-AS1 - ST7 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ST7-OT4 - ST7 overlapping transcript 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |