Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAGCGGTATGCTGTGGCCTTC[C/T]TCTGCTTGGACACGGCCTGCGTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605410 | ||||||||||||||||||||
Literature Links: |
KCND2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCND2 - potassium voltage-gated channel subfamily D member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012281.2 | 1653 | Missense Mutation | CTC,TTC | L,F 230 | NP_036413.1 | |
XM_011516165.2 | 1653 | Missense Mutation | CTC,TTC | L,F 230 | XP_011514467.1 |