Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATGCAATATTTCTGTTTATCAA[A/G]GACATCATGGATTAAATTGATAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF800 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF800 - zinc finger protein 800 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_176814.4 | 5833 | Intron | NP_789784.2 | |||
XM_005250181.3 | 5833 | Intron | XP_005250238.1 | |||
XM_006715867.3 | 5833 | UTR 3 | XP_006715930.1 | |||
XM_006715868.3 | 5833 | UTR 3 | XP_006715931.1 | |||
XM_006715869.3 | 5833 | UTR 3 | XP_006715932.1 | |||
XM_011515859.2 | 5833 | UTR 3 | XP_011514161.1 |