Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGACCGGGTGGAGCTGCCCAGCC[A/G]TACCCTCTGCCAGGCCACCCGAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SMO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SMO - smoothened, frizzled class receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005631.4 | 277 | Missense Mutation | CAT,CGT | H,R 144 | NP_005622.1 | |
XM_011516522.2 | 277 | Missense Mutation | CAT,CGT | H,R 14 | XP_011514824.1 |