Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTGCCTGCTTCTCCATGCTGCC[C/T]AGGGAGGGCCCAATATGCTGTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3HC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3HC1 - zinc finger C3HC-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282190.1 | Intron | NP_001269119.1 | ||||
NM_001282191.1 | Intron | NP_001269120.1 | ||||
NM_016478.4 | Intron | NP_057562.3 | ||||
XM_005250403.2 | Intron | XP_005250460.1 | ||||
XM_011516288.2 | Intron | XP_011514590.1 | ||||
XM_011516289.2 | Intron | XP_011514591.1 | ||||
XM_011516290.2 | Intron | XP_011514592.1 | ||||
XM_017012287.1 | Intron | XP_016867776.1 | ||||
XM_017012288.1 | Intron | XP_016867777.1 |