Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGGGAAAGCTTGGGTCTTCCCA[C/T]GACCCATCCCTCACAGAGCCTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604280 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLXNA4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLXNA4 - plexin A4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105543.1 | 12247 | Intron | NP_001099013.1 | |||
NM_020911.1 | 12247 | UTR 3 | NP_065962.1 | |||
NM_181775.3 | 12247 | Intron | NP_861440.2 | |||
XM_005250686.4 | 12247 | UTR 3 | XP_005250743.1 | |||
XM_006716171.3 | 12247 | UTR 3 | XP_006716234.1 | |||
XM_011516676.2 | 12247 | Intron | XP_011514978.1 | |||
XM_017012779.1 | 12247 | UTR 3 | XP_016868268.1 |