Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGATGGGGAGAGTGAATGCAGAAA[C/T]GGCGCATACAACACAGACAGATGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611700 | ||||||||||||||||||||
Literature Links: |
SVOPL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SVOPL - SVOP like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139456.1 | 1111 | Missense Mutation | ATT,GTT | I,V 474 | NP_001132928.1 | |
NM_174959.3 | 1111 | Missense Mutation | ATT,GTT | I,V 322 | NP_777619.1 | |
XM_005250143.3 | 1111 | Missense Mutation | ATT,GTT | I,V 474 | XP_005250200.1 | |
XM_011515797.2 | 1111 | Missense Mutation | ATT,GTT | I,V 322 | XP_011514099.1 | |
XM_017011746.1 | 1111 | Missense Mutation | ATT,GTT | I,V 383 | XP_016867235.1 | |
XM_017011747.1 | 1111 | Missense Mutation | ATT,GTT | I,V 322 | XP_016867236.1 | |
XM_017011748.1 | 1111 | Missense Mutation | ATT,GTT | I,V 322 | XP_016867237.1 |