Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGTGACTGGGCTTTGGTAAAAG[C/T]TCAGCAAGAAATGAAGAGTAGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3HAV1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3HAV1L - zinc finger CCCH-type containing, antiviral 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_080660.3 | Intron | NP_542391.2 | ||||
XM_006716176.3 | Intron | XP_006716239.1 | ||||
XM_011516688.2 | Intron | XP_011514990.1 | ||||
XM_011516689.2 | Intron | XP_011514991.1 |