Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCTTAACATTTTAGTAATTCCA[C/T]TTTCTTACTCTGATTGTTATTTTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DENND4C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DENND4C - DENN domain containing 4C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017925.6 | Intron | NP_060395.5 | ||||
XM_006716804.3 | Intron | XP_006716867.1 | ||||
XM_006716805.3 | Intron | XP_006716868.1 | ||||
XM_006716807.3 | Intron | XP_006716870.1 | ||||
XM_011517959.2 | Intron | XP_011516261.1 | ||||
XM_017014917.1 | Intron | XP_016870406.1 | ||||
XM_017014918.1 | Intron | XP_016870407.1 | ||||
XM_017014919.1 | Intron | XP_016870408.1 | ||||
XM_017014920.1 | Intron | XP_016870409.1 |