Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATCTCATCATCCAGTGAAAATTA[C/T]GCATAATAAACACAACACATACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609838 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC24A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC24A2 - solute carrier family 24 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193288.2 | 10504 | UTR 3 | NP_001180217.1 | |||
NM_020344.3 | 10504 | UTR 3 | NP_065077.1 | |||
XM_005251425.2 | 10504 | UTR 3 | XP_005251482.1 | |||
XM_005251426.4 | 10504 | UTR 3 | XP_005251483.1 | |||
XM_006716750.3 | 10504 | UTR 3 | XP_006716813.1 | |||
XM_017014592.1 | 10504 | UTR 3 | XP_016870081.1 | |||
XM_017014593.1 | 10504 | UTR 3 | XP_016870082.1 |